Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber, GLS 80™ Thread, USP /EP (3.2.1) Light Transmission, Graduated
Wide mouth, multifunctional glass laboratory bottles with UV protection.
£31.41 - £870.00
Specifications
For Use With (Application) | Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir |
---|---|
Includes | Without screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15971371
|
DWK Life Sciences
1178429 |
500 mL |
£485.00
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
15981371
|
DWK Life Sciences
1178430 |
1000 mL |
£595.00
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
15991371
|
DWK Life Sciences
1178431 |
2000 mL |
£870.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
10163003
|
DWK Life Sciences
1178433 |
10,000 mL |
£423.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10657053
|
DWK Life Sciences
1178434 |
20,000 mL |
£695.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10777214
|
DWK Life Sciences
1178432 |
5000 mL |
£279.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
12676167
|
DWK Life Sciences
218663604 |
250 mL |
£445.00
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
10647053
|
DWK Life Sciences
1178431 |
2000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10796444
|
DWK Life Sciences
1178430 |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10669353
|
DWK Life Sciences
1178429 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11639938
|
DWK Life Sciences
218666901 |
3500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir | |
Borosilicate Glass (3.3) | |
Autoclavable |
Without screw cap and pouring ring from PP (blue) | |
GLS 80 | |
Polypropylene |