missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Dulcitol, 99+%
£25.83 - £197.99
Chemical Identifiers
CAS | 608-66-2 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
MDL Number | MFCD00064288 |
InChI Key | FBPFZTCFMRRESA-GUCUJZIJSA-N |
Synonym | dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol |
PubChem CID | 11850 |
ChEBI | CHEBI:16813 |
IUPAC Name | (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol |
SMILES | C(C(C(C(C(CO)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10279213
|
Thermo Scientific Chemicals
117700250 |
25 g | Glass bottle |
£25.83
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10569430
|
Thermo Scientific Chemicals
117701000 |
100 g | Glass bottle |
£47.13
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10608254
|
Thermo Scientific Chemicals
117705000 |
500 g | Glass bottle |
£197.99
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
608-66-2 | |
182.17 | |
FBPFZTCFMRRESA-GUCUJZIJSA-N | |
11850 | |
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
MFCD00064288 | |
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol | |
CHEBI:16813 | |
C(C(C(C(C(CO)O)O)O)O)O |
Specifications
608-66-2 | |
White | |
Authentic | |
C6H14O6 | |
01, 544 | |
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol | |
FBPFZTCFMRRESA-GUCUJZIJSA-N | |
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol | |
11850 | |
182.17 | |
Powder |
188°C to 189°C | |
275°C to 280°C (1.0 mmHg) | |
99% min. (ELSD) (HPLC) | |
MFCD00064288 | |
15, 4362 | |
(3% in water) Colorless | |
C(C(C(C(C(CO)O)O)O)O)O | |
182.17 | |
CHEBI:16813 | |
99+% | |
Dulcitol, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 210-165-2
missing translation for 'tsca' : TSCA
Product Identifier
- Dulcitol
RUO – Research Use Only