Learn More
Applied Biosystems™ DS-33 Matrix Standard Kit Dx (Dye Set G5)
Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples.
Brand:  Applied Biosystems™ A25775
Product Code. 15998826
  • £214.00 / 8 tests
Explore more special offers
Includes: Contains 5-dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution
Description
Description
The DS-33 Matrix Standard Kit Dx (Dye Set G5) is used to generate the 'multicomponent matrix' required when analyzing 6-FAM™, VIC™, NED™, PET™, and LIZ™ dye-labeled DNA fragments on the Applied Biosystems™ 3500 Dx Series Genetic Analyzers.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains 5-dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution. Store at 2 to 8°C
3500 Dx Genetic Analyzer
DS-33 Matrix Standard Kit
Dx DS-33 Matrix Standard Kit, 8 runs, LIZ™ Dye, Wet Ice, For 3500 Dx Genetic Analyzer, 2 to 8°C Storage Temperature
LIZ
8 runs