missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Dot Blot Hybridisation Manifold System
Precision lapped mating surfaces that ensures uniform filter/membrane contact.
£814.00 - £927.00
Specifications
Type | Dot Blot Hybridization System |
---|---|
Temperature (Metric) Operating | 65°C |
Volume (Metric) Sample | 100 μL |
Dimensions (L x W x H) | D x W x H: 7.5 x 18.5 x 7 cm |
Product Code | Brand | Device Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Device Holds | Price | Quantity & Availability | |||||
11837172
|
Fisherbrand™
|
48 Samples |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11799545
|
Fisherbrand™
|
96 Samples |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High density acrylic
- Microfiltration manifold
- Allows unfractionated samples to be immobilised onto membrane
- Immediate screening with nucleic acid probes or antibodies
- Upper manifold - allows sample to be loaded into each dot/slot
- Stainless steel thumbscrews and precision-lapped mating surfaces
- Template to allow membrane to be cut down to size
- Vacuum Port, can be connected to water aspirator or vacuum pump
- Vacuum reservoir to serve as collection chamber for filtrate
- Requires vacuum pump or water aspirator equivalent to 600mm Hg (0.8 bar)
- DHM-48: Used for 5 x 12.8cm membrane - 48 sample throughput
- DHM-96: Used for 7.5 x 11.3cm membrane - 96 sample throughput
- Compatible with multichannel pipettes and standard 96-well microtitre plates
Specifications
Dot Blot Hybridization System | |
65°C | |
100 μL | |
D x W x H: 7.5 x 18.5 x 7 cm |