missing translation for 'onlineSavingsMsg'
Learn More

Domiphen bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7046051
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
538-71-6
C22H40BrNO
414.472
MFCD00011769
OJIYIVCMRYCWSE-UHFFFAOYSA-M
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
10866
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]
This item is not returnable. View return policy
538-71-6
C22H40BrNO
414.472
MFCD00011769
OJIYIVCMRYCWSE-UHFFFAOYSA-M
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
10866
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 117°C to 118°C
Quantity 25 g
Beilstein 3922742
Sensitivity Hygroscopic
Merck Index 14,3415
Solubility Information Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene.
Formula Weight 414.48
Percent Purity 97%
Chemical Name or Material Domiphen bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'