missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%
£57.69 - £186.93
Chemical Identifiers
CAS | 69833-18-7 |
---|---|
Molecular Formula | C4H8N2O3 |
Molecular Weight (g/mol) | 132.12 |
MDL Number | MFCD00151039,MFCD00008036,MFCD00064400 |
InChI Key | DCXYFEDJOCDNAF-UHFFFAOYNA-N |
Synonym | dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina |
PubChem CID | 522362 |
SMILES | NC(CC(N)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11469766
|
Thermo Scientific Chemicals
B20273.22 |
100 g |
£57.69
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479766
|
Thermo Scientific Chemicals
B20273.36 |
500 g |
£186.93
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.
Solubility
Soluble in water, acids, and alkalis.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
69833-18-7 | |
132.12 | |
DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
522362 |
C4H8N2O3 | |
MFCD00151039,MFCD00008036,MFCD00064400 | |
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
NC(CC(N)=O)C(O)=O |
Specifications
∼220°C (decomposition) | |
C4H8N2O3 | |
6234992 | |
Soluble in water,acids,and alkalis. | |
NC(CC(N)=O)C(O)=O | |
132.12 | |
150.14 (132.12 Anhydrous) | |
DL-Asparagine monohydrate |
69833-18-7 | |
MFCD00151039,MFCD00008036,MFCD00064400 | |
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
2-amino-3-carbamoylpropanoic acid | |
522362 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 221-521-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only