missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Divided Storage Tray

Divided trays are designed to be placed within a horizontal/ vertical cold store or be free standing

Brand:  Wesbart™ WB05/20

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 15888012

  • £39.30 / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

  • Use the division guide on the inside cover to check the size of the test tube you wish to store
  • As the cells have no spacing in between remember to accommodate for the cap or lid size too
  • Specifications

    Specifications

    Divided Tray
    6 x 14
    307 mm
    128 mm
    20 mm dividers
    Samples (84)
    34 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers