missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Divided Storage Tray

Divided trays are designed to be placed within a horizontal/ vertical cold store or be free standing

Brand:  Wesbart™ WB08/16

Code : A0

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 11746324

  • £16.90 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Use the division guide on the inside cover to check the size of the test tube you wish to store
  • As the cells have no spacing in between remember to accommodate for the cap or lid size too
  • Specifications

    Specifications

    Divided Tray
    9 x 9
    154 x 154 x 34 mm
    34 mm
    16 mm dividers
    Samples (81)
    154 mm
    154 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers