missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disuccinimidyl glutarate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Description
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.
Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.
Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Quantity | 100 mg |
Beilstein | 8575145 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water. |
Formula Weight | 326.26 |
Percent Purity | 97% |
Chemical Name or Material | Disuccinimidyl glutarate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'