missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, Multi-Compendial, J.T. Baker™
378.10 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Specifications
Quantity | 100 mL |
Flash Point | 89.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.10kg |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13 |
Grade | Multi-Compendial |
Chemical Name or Material | Dimethyl Sulfoxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'