missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3735850
Click to view available options
Quantity:
2.5 mL
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
381-73-7
C2H2F2O2
96.03
MFCD00004220
PBWZKZYHONABLN-UHFFFAOYSA-N
difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
9788
CHEBI:23716
2,2-difluoroacetic acid
C(C(=O)O)(F)F
This item is not returnable. View return policy
381-73-7
C2H2F2O2
96.03
MFCD00004220
PBWZKZYHONABLN-UHFFFAOYSA-N
difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
9788
CHEBI:23716
2,2-difluoroacetic acid
C(C(=O)O)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -1°C
Color Colorless to Brown
Density 1.5200g/mL
Boiling Point 132°C to 134°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2CHCO2H
Refractive Index 1.3420 to 1.345
Quantity 2.5 mL
Beilstein 02, 193
Specific Gravity 1.52
Formula Weight 96.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Difluoroacetic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'