missing translation for 'onlineSavingsMsg'
Learn More

Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-109062
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
1470-61-7
C5H10AgNS2
256.13
MFCD00004929
NSVHDIYWJVLAGH-UHFFFAOYSA-M
silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
3034078
silver;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].[Ag+]
1470-61-7
C5H10AgNS2
256.13
MFCD00004929
NSVHDIYWJVLAGH-UHFFFAOYSA-M
silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
3034078
silver;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172.0°C to 175.0°C
Color Khaki
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C2H5)2NCS2Ag
Quantity 10 g
Beilstein 04,121
Solubility Information (in Pyridine) Passes test
Formula Weight 256.13
Grade ACS Reagent
Physical Form Amorphous Powder
Chemical Name or Material Diethyldithiocarbamic acid, silver salt
Show More Show Less
compliance-icons
Product Identifier
  • Diethyldithiocarbamic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'