missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Anhydrous, BAKER ANALYZED (ACS), J.T. Baker™

Product Code. 10231201
Click to view available options
Quantity:
1 L
5 L
Packaging:
Aluminum Bottle
SAFET
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use. Stabilized with 7ppm BHT.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.3°C
Color Colorless
Density 0.71g/cm3 at 20°C
Boiling Point 35°C
Quantity 5 L
Explosion Limit 1.7 - 48 Vol %
Flash Point −40°C
Ignition Point 170°C
Odor Sweetish
Packaging Aluminum Bottle
Physical Form Fluid
Residue after Evaporation Max. 0.001%
Titratable Acid Max. 0.0002meq/g
Linear Formula (C2H5)2O
Viscosity 0.233 mPass at 20°C
Water Max. 0.01%
Solubility Information 12g/L in water
Formula Weight 74.12g/mol
Percent Purity ≥99.5%
Grade Exceeds ACS specifications
Chemical Name or Material Diethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl Ether, Anhydrous Baker Analyzed ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Diethyl ether
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'