missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.9+%, for Nucleotide and Peptide Synthesis, Stabilised with Amylene, Fisher Chemical™
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
ClCCl |
CH2Cl2 | |
MFCD00000881 | |
dichloromethane |
Specifications
-97°C | |
Colorless | |
Liquid | |
MFCD00000881 | |
0.43 mPaS at 20°C | |
ClCCl | |
84.93 |
75-09-2 | |
39°C | |
CH2Cl2 | |
350mbar at 20°C | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
84.93g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dichloromethane, 99.9+%, for Nucleotide and Peptide Synthesis, Stabilised with Amylene, Fisher Chemical™