Learn More
Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II), 98%, Thermo Scientific Chemicals
Description
Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II) is a reactant involved in Kumada catalyst-transfer polycondensation, oxidation of carboranyl phosphine ligands, synthesis of nickel-iron dithiolato hydrides, Ullmann reactions - homocoupling reactions, co-catalyst for hydroformylation of alcohols for the synthesis of quaternary carbon centers, catalyst for Wacker-type intramolecular aerobic oxidative amination of alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 263°C to 265°C |
Color | Brown to Orange-Red |
Quantity | 2 g |
Solubility Information | Insoluble in water. |
Formula Weight | 528.02 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II) |
RUO – Research Use Only