missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetonitrile, 94%, Thermo Scientific Chemicals

Product Code. 11478836
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3252-43-5
Molecular Formula C2HBr2N
Molecular Weight (g/mol) 198.845
MDL Number MFCD00001856
InChI Key NDSBDLSWTGLNQA-UHFFFAOYSA-N
Synonym dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940
PubChem CID 18617
ChEBI CHEBI:82445
IUPAC Name 2,2-dibromoacetonitrile
SMILES C(#N)C(Br)Br

Specifications

Density 2.296
Boiling Point 67°C to 69°C (24 mmHg)
Flash Point 56°C (133°F)
Odor Pungent, Makes Eyes Water
Linear Formula Br2CHCN
Refractive Index 1.542
Quantity 100 g
UN Number UN3286
Beilstein 1739037
Sensitivity Light sensitive
Formula Weight 198.84
Percent Purity 94%
Chemical Name or Material Dibromoacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'