Learn More
DHRS11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561691
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EGLRQELREA QTHIRATCIS PGVVETQFAF KLHDKDPEKA AATYEQMKCL KPEDVAEAVI YVLSTPAHIQ IGDIQMRPTE QVT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.Specifications
DHRS11 | |
Polyclonal | |
Unconjugated | |
DHRS11 | |
17-beta-hydroxysteroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; ARPG836; dehydrogenase/reductase (SDR family) member 11; dehydrogenase/reductase 11; dehydrogenase/reductase SDR family member 11; DHRS11; Estradiol 17-beta-dehydrogenase; hypothetical protein LOC510264; inactive dehydrogenase/reductase isoform; LOW QUALITY PROTEIN: dehydrogenase/reductase SDR family member 11; RGD1307935; SDR24C1; short chain dehydrogenase/reductase family 24C, member 1; short-chain dehydrogenase/reductase family 24C member 1; spDHRS11; UNQ836/PRO1774 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWP2 | |
DHRS11 | |
Recombinant protein corresponding to Human DHRS11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only