Learn More
DGKA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565409
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: KVSDVLKLFED GEMAKYVQGD AIGYEGFQQF LKIYLEVDNV PRHLSLALFQ SFETGHCLNE TNVTKDVVCL NDVSCYFSL Highest antigen sequence identity to the following orthologs - mouse 58%, rat 58%.
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Specifications
DGKA | |
Polyclonal | |
Unconjugated | |
Dgka | |
80 kDa diacylglycerol kinase; 80kDa; AW146112; DAG kinase alpha; Dagk; Dagk1; Dgka; DGK-alpha; diacylglycerol kinase; diacylglycerol kinase alpha; diacylglycerol kinase alpha Flt2-Pr2; diacylglycerol kinase, alpha; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase; diglyceride kinase alpha; MGC12821; MGC42356; truncated diacylglycerol kinase alpha Flt1-Pr1; truncated diacylglycerol kinase alpha Flt1-Pr2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1606 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23743 | |
Dgka | |
Recombinant Human DGKA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only