Learn More
Cyclopentanone-2,2,5,5-d{4}, 95%, Thermo Scientific Chemicals
£236.00 - £772.00
Chemical Identifiers
CAS | 3997-89-5 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 88.142 |
MDL Number | MFCD00190415 |
InChI Key | BGTOWKSIORTVQH-KHORGVISSA-N |
Synonym | cyclopentanone-2,2,5,5-d4, cyclopentanone d4, 2,2,5,5-∼2∼h_4_ cyclopentanone, cyclopentanone-2,2,5,5-d4, 98 atom % d, 2,2,5,5-2 h? cyclopentan-1-one |
PubChem CID | 16213326 |
IUPAC Name | 2,2,5,5-tetradeuteriocyclopentan-1-one |
SMILES | C1CCC(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457977
|
Thermo Scientific Alfa Aesar
H51095.03 |
1 g |
£236.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15467977
|
Thermo Scientific Alfa Aesar
H51095.06 |
5 g |
£772.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3997-89-5 | |
88.142 | |
BGTOWKSIORTVQH-KHORGVISSA-N | |
16213326 | |
C1CCC(=O)C1 |
C5H8O | |
MFCD00190415 | |
cyclopentanone-2,2,5,5-d4, cyclopentanone d4, 2,2,5,5-∼2∼h_4_ cyclopentanone, cyclopentanone-2,2,5,5-d4, 98 atom % d, 2,2,5,5-2 h? cyclopentan-1-one | |
2,2,5,5-tetradeuteriocyclopentan-1-one |
Specifications
3997-89-5 | |
MFCD00190415 | |
cyclopentanone-2,2,5,5-d4, cyclopentanone d4, 2,2,5,5-∼2∼h_4_ cyclopentanone, cyclopentanone-2,2,5,5-d4, 98 atom % d, 2,2,5,5-2 h? cyclopentan-1-one | |
C1CCC(=O)C1 | |
88.142 | |
88.14 | |
Cyclopentanone-2,2,5,5-d4 |
C5H8O | |
UN2245 | |
BGTOWKSIORTVQH-KHORGVISSA-N | |
2,2,5,5-tetradeuteriocyclopentan-1-one | |
16213326 | |
95% |
Safety and Handling
GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANONE
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only