missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CXCL1 (GROα) Recombinant Human Protein
CXCL1 (GROα) Recombinant Human Protein
Brand: Gibco™ PHC1066
Code : 50
Additional Details : Weight : 0.50000kg
Description
Recombinant Human Chemokine (C-X-C motif) ligand 1 (Also known as CXCL1, GROα, melanoma growth stimulating activity, MSGA, Neutrophil-activating protein 3, NAP-3) is a bioactive protein intended for use in cell culture applications. CXCL1 is a chemokine that induces the infiltration of neutrophils.High Purity Recombinant Bioactive Protein
Recombinant Human Chemokine (C-X-C motif) ligand 1 is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Chemokine (C-X-C motif) ligand 1 (CXCL1, GROα) specific activity is determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
See our selection of recombinant Gibco™ growth factors.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Specifications
Cell Culture | |
2919 | |
CXCL1 (GROα±) Recombinant Human Protein | |
25μg | |
For research use only. Not for use in diagnostic procedures | |
CXCL1 (GROa) | |
The biological activity was determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils. Significant calcium mobilization is observed with ≥1ng/mL of recombinant human GRO-alpha. The optimal concentration for each specific application should be determined by an initial dose response assay | |
Human | |
Chemokines and receptors | |
GRO/KC/CINC Chemokines |
Lyophilized | |
8kDa | |
Sequential chromatography | |
2 to 8°C | |
1 Each | |
CXCL1 (GROα±) Recombinant Human Protein | |
CXCL1 (GROα±) Recombinant Human Protein | |
E. coli | |
Lyophilized | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.