Learn More
Thermo Scientific™ Culti-Loops™ Streptomyces griseus subsp. griseus ATCC™ 10137™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4609013
Code : 65
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
10137TM* | |
Culti-Loops | |
Gram-positive Rods | |
5 Loops/Pack |
Streptomyces griseus subsp. griseus ATCC™ 10137™ | |
Culti-Loop | |
Qualitative QC | |
4 [BUCSAV 7, Boots FD40, NCIB 8232, NCTC 6961] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.