Learn More
Thermo Scientific™ Culti-Loops™ Bacteroides thetaiotaomicron ATCC™ 29741™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4601260
193.17 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Bacteroides species | |
Bacteroides thetaiotaomicron | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
WAL 2926 [CIP 104207] |
29741TM* | |
Culti-Loops | |
Anaerobes | |
5 Loops/Pack |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.