missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK1G1 (CK1 γ 1) Recombinant Human Protein
CSNK1G1 (CK1 gamma 1) is one of the most abundant serine/threonine kinases in eukaryotic cell extracts.
£1602.00 - £6900.00
Specifications
Accession Number | NP_071331 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 72.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408583
|
Invitrogen™
PR7391A |
100μg |
£1602.00
100µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10463843
|
Invitrogen™
PV3989 |
1mg |
£6900.00
1mg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Multiple isoforms of the CSNK1 enzyme exist, and the gamma-1 isoform is involved in growth and morphogenesis of eukaryotic cells.
Specifications
NP_071331 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
72.9kDa | |
RUO - research use only | |
CSNK1G1 (CK1 γ 1) | |
Human | |
GST Tag | |
Frozen |