missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1E (CK1 ε) Recombinant Human Protein
CSNK1E (CK1 ε) Recombinant Human Protein
Brand: Invitrogen PV3500
Code : 50
Additional Details : Weight : 0.00945kg
Description
CSNK1E (CK1 epsilon) is a serine⁄threonine kinase implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis.Specifications
NP_001885 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
52.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
CSNK1E (CK1 e) | |
CSNK1E (CK1 epsilon) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 420μg/mL dephosphorylated casein substrate and trace [ 32P]-γ-ATP for 10 minutes at 30°C | |
CSNK1E (CK1 αμ) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.42mg/mL | |
1454 | |
CSNK1E (CK1 αμ) Recombinant Human Protein | |
–80°C | |
1 Each | |
CSNK1E (CK1 αμ) Recombinant Human Protein | |
CK1 | |
460nmoles of phosphate transferred to dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.