missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK1D (CK1 δ) Recombinant Human Protein
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway.
Brand: Invitrogen™ PV3987
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The Wnt/beta-catenin signaling pathway is important in both development and cancer.
Specifications
NP_620693 | |
0mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
CSNK1D (CK1 δ) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |