Learn More
Cole-Parmer™ CPC™ Panel-mount Gasket for Miniature Non-spill Connectors
With the miniature non-spill quick-disconnect coupling, fluid line protection has never been easier
Brand:  Cole-Parmer™ 31604-61
Product Code. 15487393
Description
Description
  • Less than 0.025mL of spillage.
  • Unique valve design for ultra-low air inclusion in line.
  • Non-spill protection and high-flow fluid processing with a small footprint.
  • Economical alternative for non-spill solutions.
  • This coupling features an innovative twist-to-connect design with an integrated locking mechanism for secure coupling engagement and worry-free fit.
  • Both halves of the coupling include shut-off valves to guarantee minimal spillage when disconnected.
  • The same shut-off valves ensure less than 0.009mL air inclusion to minimize contaminants in the process line.
  • This slim, compact coupling reduces valuable space required for the connection and minimizes components in your process.
Specifications
Specifications
Connector
10/Pk.
Quick-Disconnect
Buna N
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ CPC™ Panel-mount Gasket for Miniature Non-spill Connectors > Material: Buna N