missing translation for 'onlineSavingsMsg'
Learn More

COXIV Mouse anti-Human, Clone: 2A7B2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66110-1-IG-150UL

Additional Details : Weight : 0.00010kg

Product Code. 16877685

  • £189.00 / 150µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to COX4I1 and do not cross reacts with COX4I2.

Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications

Specifications

COXIV
Monoclonal
1.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13073
COX4I1
COXIV Fusion Protein Ag20551
150 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2A7B2
Unconjugated
COX4I1
COX IV 1, COX4, COX4I1, COXIV, Cytochrome c oxidase polypeptide IV, Cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Mouse
Protein A
RUO
1327
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers