Learn More
Corning™ Costar™ Stripettor™ Pipet Filler/Dispenser Replacement Parts
For use with Stripettor™ filler/dispenser
Brand: Corning™ 4096
Code : NEW
Additional Details : Weight : 0.04990kg
Description
- PTFE filter (pack of 5) is for the Stripettor™ Plus pipetting controller
- Pore Size: 0.45 μm
- Autoclavable
Specifications
Not Autoclavable | |
Silicone | |
Pipet Holder | |
Costar™ stripettor pipet filler/dispenser |