Learn More
Corning™ Block
Material: Stainless Steel; For Use With: LSE Digital Dry Bath Heater
Brand: Corning™ 480131
Code : 04
Additional Details : Weight : 0.81647kg
Description
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
Specifications
LSE Digital Dry Bath Heater | |
Stainless Steel | |
Block | |
96-well plate or 4 slides |