missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper phthalocyanine, dye content ca 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Merck Index | 15, 2505 |
Solubility Information | Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons |
Infrared Spectrum | Authentic |
Formula Weight | 576.08 |
Percent Purity | ≥94% (ex Cu) |
Packaging | Glass bottle |
UV Spectrometry | Complies |
Color | Blue |
Quantity | 25 g |
Physical Form | Fine Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'