missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 1
PCA and TTC and Neutralizing / VRBG Agar and Neutralizing, Flex Dip-slide with a selective medium for detection of Enterobacteriaceae and a non selective medium for total bacterial count.
POA
Specifications
Species | Escherichia coli, Proteus species, Klebsiella species, Pseudomonas species |
---|---|
Certifications/Compliance | ISO 18593:2004 |
Content And Storage | 10°C to 25°C |
Detects | Bacterial Count, Enterobacteriaceae |
Format | Slide |
Description
- Contact Slide 1 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of Enterobacteriaceae . The other medium is used to obtain the total bacterial count.
Specifications
Escherichia coli, Proteus species, Klebsiella species, Pseudomonas species | |
10°C to 25°C | |
Slide | |
Contact Slide | |
PCA, TTC, Neutralizing/VRBG Agar, Neutralizing | |
At 36 ± 1°C for 24 to 48 hr. | |
ISO 18593:2004 |
ISO 18593:2004 | |
Bacterial Count, Enterobacteriaceae | |
Italy | |
Non-sterile | |
For Bacterial Count and Enterobacteriaceae Detection | |
24 to 48hr. | |
Flex |