Learn More
Fisherbrand™ Combs for omniPAGE Mini Vertical Gel Electrophoresis Unit, 0.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£15.75
Product Code | Brand | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | ||||
11857583
|
Fisherbrand™
|
10 | 20 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11837593
|
Fisherbrand™
|
12 | 16 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11887593
|
Fisherbrand™
|
16 | 12.5 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11837603
|
Fisherbrand™
|
20 | 10 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11887603
|
Fisherbrand™
|
5 | 50 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11837613
|
Fisherbrand™
|
8 | 30 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11887613
|
Fisherbrand™
|
9 | 25 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11837583
|
Fisherbrand™
|
1 Prep, 1 Marker | 330 μL |
£15.75
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
Description
- Accessory for use with omniPAGE mini vertical gel electrophoresis unit
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Comb | |
0.5 mm |