Learn More
Epredia™ ColorStack Storage System
Take organization of your slides archives one step further with color-coded drawers.
Brand:  Epredia™ E43/12C/TAN
Code : 50
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12111830
  • £331.00 / Each
Estimated Shipment: 30-07-2024
to see stock.
Description
Description
  • Fully compatible with all standard cabinets, bases, and options
  • Cabinet is cream epoxy-coated metal; drawers have colored plastic fronts
  • Each cabinet comes with 14 drawers of one color
  • Drawers in the standard 14-Drawer Cabinet can easily be switched out with the ColorStack drawers
  • Special top cover is cream to match the ColorStack 14-Drawer cabinet color
Specifications
Specifications
14-Drawer Cabinet
Tan