Learn More
CMS™ Water Vapor Chips
Brand: Draeger Safety 6406450
Code : 50
Additional Details : Weight : 0.02880kg
Specifications
700 to 1100 hpa | |
0.4 to 10ppm | |
0°C to 40°C |
Water Vapor Chip | |
20 to 150sec. |