Learn More
Invitrogen™ CLK4 Recombinant Human Protein
CLK4 belongs to the CDC2-like protein kinase (CLK) family.
Brand: Invitrogen™ PR7453A
Code : NEW
Additional Details : Weight : 0.00900kg
Description
CLK4 can interact with and phosphorylate the serine- and arginine-rich (SR) proteins, which are known to play an important role in the formation of spliceosomes, and thus is thought to be involved in the regulation of alternative splicing.
Specifications
NP_065717 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CLK4 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
85kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CLK Kinase Family | |
Recombinant | |
Baculovirus |