Learn More
DWK Life Sciences Cleaning Scraper, for glass ceramic laboratory protection plate
Ideal for cleaning glass ceramic plates.
Brand:  DWK Life Sciences 290790109
Code : NEW
Additional Details : Weight : 0.68000kg
Product Code. 15282109
  • £196.00 / Pack of 10
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • The blade allows easy cleaning of the glass ceramic plate
Specifications
Specifications
Not Autoclavable
Cleaning of glass ceramic plates
10/Pk.