Learn More
Choline iodide, 98%, Thermo Scientific Chemicals
£25.40 - £247.00
Chemical Identifiers
CAS | 17773-10-3 |
---|---|
Molecular Formula | C5H14INO |
Molecular Weight (g/mol) | 231.08 |
MDL Number | MFCD00011900 |
InChI Key | FNPBHXSBDADRBT-UHFFFAOYSA-M |
Synonym | choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide |
PubChem CID | 87300 |
IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;iodide |
SMILES | [I-].C[N+](C)(C)CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314334
|
Thermo Scientific Alfa Aesar
L00860.09 |
10 g |
£25.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324334
|
Thermo Scientific Alfa Aesar
L00860.18 |
50 g |
£76.90
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334334
|
Thermo Scientific Alfa Aesar
L00860.30 |
250 g |
£247.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17773-10-3 | |
231.08 | |
FNPBHXSBDADRBT-UHFFFAOYSA-M | |
87300 | |
[I-].C[N+](C)(C)CCO |
C5H14INO | |
MFCD00011900 | |
choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide | |
2-hydroxyethyl(trimethyl)azanium;iodide |
Specifications
17773-10-3 | |
C5H14INO | |
3912691 | |
FNPBHXSBDADRBT-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;iodide | |
87300 | |
98% |
∼270°C | |
MFCD00011900 | |
choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide | |
[I-].C[N+](C)(C)CCO | |
231.08 | |
231.08 | |
Choline iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'rtecsNumber' : GA4100000
RUO – Research Use Only