missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Purified, Macron Fine Chemicals™
185.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- No current compendia
- Formerly listed in the USP and NF compendium
Specifications
Melting Point | -63.5°C |
CAS | 67-66-3 |
Color | Colorless |
Molecular Formula | CHCl3 |
MDL Number | MFCD00000826 |
Appearance | Clear |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
SMILES | ClC(Cl)Cl |
IUPAC Name | trichloromethane |
Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'