missing translation for 'onlineSavingsMsg'
Learn More

CHIR 99021, Tocris Bioscience™

Catalog Number p-4816967
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
252917-06-9
C22H18Cl2N8
465.34
MFCD11846251
AQGNHMOJWBZFQQ-UHFFFAOYSA-N
gsk-3 inhibitor xvi, unii-234cmt4gk4, chir-911, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-ylamino ethylamino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile
9956119
6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile
CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1
This item is not returnable. View return policy
252917-06-9
C22H18Cl2N8
465.34
MFCD11846251
AQGNHMOJWBZFQQ-UHFFFAOYSA-N
gsk-3 inhibitor xvi, unii-234cmt4gk4, chir-911, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-ylamino ethylamino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile
9956119
6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile
CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1

Highly selective GSK-3 inhibitor

CHIR 99021 is a potent and highly selective inhibitor of glycogen synthase kinase 3 (GSK-3) (IC50 values are 6.7 and 10 nM for GSK-3β and GSK-3α respectively). Acts as Wnt activator. Exhibits >500-fold selectivity for GSK-3 over closely related kinases; also displays >800-fold selectivity against 45 additional enzymes and receptors. In combination with the irreversible inhibitor of LSD1 (Cat. No. 3852), CHIR 99021 enables reprogramming of mouse embryonic fibroblasts, transduced by Oct4 and Klf4 only, into iPSCs. Enhances mouse and human ESC self-renewal when used in combination with PD 0325901 (Cat. No. 4192). It promotes differentiation of hPSCs in cardiomyocytes and allows formation of extended pluripotent stem (EPS) cells in combination with (S)-(+)-Dimethindene maleate (Cat. No. 1425), tetracycline antibiotic (Cat. No. 3268) and human leukemia inhibitory factor. CHIR 99021 synthesized to cGMP guidelines also available. For more information on how to use CHIR 99021, see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts .
Quantity 10 mg
Formula Weight 465.34
Percent Purity >99%
Chemical Name or Material CHIR 99021
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'