missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, Reagent Grade, 99.9%, Honeywell™
Click to view available options
Quantity:
25 g
100 g
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 100 g |
Linear Formula | CsCl |
UN Number | NONH for all modes of transport |
Formula Weight | 168.36g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Packaging | Poly bottle |
Chemical Name or Material | Cesium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'