Learn More
Cytiva Celtron Filter Holder
Brand: Cytiva 10462270
Code : NEW
Additional Details : Weight : 2.18000kg
Specifications
Celtron 30/0.45, CN-GF51 Filter | |
Cellulose acetate membrane, polypropylene housing | |
0.45 μm | |
Difficult-to-filter solutions |