missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDK1/Cyclin B, His Tag Recombinant Protein
Human CDK1/Cyclin B, His Tag Recombinant Protein
Brand: Invitrogen™ PV3980
Code : 50
Additional Details : Weight : 0.05250kg
Description
CDK1 is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF). It is essential for G1⁄S and G2⁄M phase transitions of the eukaryotic cell cycle, and plays important regulatory roles in cell cycle control. This protein is coexpressed and copurified with CyclinB.Specifications
NP_001777 I NP_114172 | |
Protein Assays and Analysis | |
91kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
CDK1/Cyclin B | |
CDK1/cyclin B was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 5mM MgCl2, 100 μM ATP, 100μg/mL Histone H1 peptide substrate (PKTPKKAKKL) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
CDK1/Cyclin B Recombinant Human Protein | |
Human | |
Baculovirus | |
80% by SDS-PAGE |
0.16mg/mL | |
Frozen | |
CDK1/Cyclin B Recombinant Human Protein | |
–80°C | |
1 Each | |
CDK1/Cyclin B Recombinant Human Protein | |
CDK and CHK kinase family | |
1540nmoles of phosphate transferred to histone H1 peptide substrate (PKTPKKAKKL) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5μg/mL | |
His Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.