Learn More
IBA Lifesciences CD19 Isolation Kits for FABian™, Human
IBA Lifesciences CD19 Isolation Kits for FABian™, Human for reversible cell isolation delivering label-free target cells.
Brand: IBA Lifesciences 6-6013-001
Additional Details : Weight : 0.11200kg
Description
- The kit is used for the selection of CD19+ cells from whole blood or buffy coat in conjunction with the FABian™ system
- It includes CD19 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe
- Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood or 6.4 mL buffy coat
Specifications
Cell Isolation Kit | |
Selection of CD19+ Cells From Whole Blood or Buffy Coat in Conjunction with The FABian™ System | |
1 Column |
2°C to 8°C | |
1 Kit |