missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Caspase-1 Inhibitor I
£66.20 - £230.00
Chemical Identifiers
CAS | 143313-51-3 |
---|---|
Molecular Formula | C23H32N4O8 |
Synonym | Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406369
|
Thermo Scientific Chemicals
J64407.M^ |
1 mg |
£66.20
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496359
|
Thermo Scientific Chemicals
J64407.#0 |
5 mg |
£230.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A potent, specific, and reversible inhibitor of caspases-1, -4 and -5
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
143313-51-3 | |
C23H32N4O8 | |
Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO |
Specifications
White | |
A potent, specific, and reversible inhibitor of caspases-1, -4 and -5 | |
C23H32N4O8 | |
Ac-YVAD-CHO; Ac-Tyr-Val-Ala-Asp-CHO | |
Powder | |
CC(C)[C@H](NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(=O)[C@H](C)N[C@@H](CC(O)=O)C=O | |
492.53 |
-30°C to -10°C | |
143313-51-3 | |
492.52 | |
Moisture Sensitive | |
KQCBPHUOVBKBPQ-SJVNDZIOSA-N | |
(3S)-3-{[(2S)-1-[(2S)-2-[(2S)-2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido]-1-oxopropan-2-yl]amino}-4-oxobutanoic acid |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only