missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Campingaz™ CV360 Butane Gas Cartridge
Take along wherever a heat fuel source is needed such as portable Bunsen Burners or outdoor stoves. Azpack™ Campingaz™ CV360 Butane Gas Cartridge contains 115mL of pressurized butane.
Brand: Azpack™ 12336247
This item is not returnable.
View return policy
Description
- Contains 115mL butane gas
- Weight: 52g
- Made in France
- Complies with EN417
Specifications
Gas Cartridge | |
37 mm | |
140 mm | |
6/Pk |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Azpack Campingaz CV360 Butane Gas Cartridge
Signal Word
- Danger
Hazard Category
- Flammable gas.
Hazard Statement
- H220-Extremely flammable gas.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
- P381-Eliminate all ignition sources if safe to do so.
- P403-Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : hydrocarbons C3-C4 petroleum gas