Learn More
Invitrogen™ CAR Human Protein, Ligand Binding Domain, GST-tagged
Constitutive Active Receptor, Ligand Binding Domain (CAR-LBD) is a 58.6 kDa recombinant human protein (amino acids 85-348) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand:  Invitrogen™ PV4838
Code : 50
Additional Details : Weight : 0.50000kg
Product Code. 10469813
  • £1554.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
CAR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Specifications
Nuclear Receptor Assay
Store in ultra-cold freezer (-68 to -85°C).
Ligand Binding Domain
Human
Hormone Receptors
50 μg
RUO - research use only
CAR LBD
Recombinant
Product Suggestions