missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium nitrate tetrahydrate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents.
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents.
Specifications
Specifications
Boiling Point | 132°C (decomposition) |
Melting Point | 43°C to 45°C |
Quantity | 2500 g |
UN Number | UN1454 |
Merck Index | 14,1682 |
Solubility Information | Soluble in water,alcohol and acetone. |
Formula Weight | 236.15 (164.09 Anhydrous) |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Product Identifier
- Calcium nitrate, tetrahydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'