missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride 0.02 M (0.04 N) Analytical Volumetric Solution (AVL), Reagecon
Fully factorized, high purity, stable, certified product, developed and tested for complexometric titrations.
Brand: Reagecon CACL20021
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Complexometric titrimetry is based on the reaction of a metal ion with a ligand to form a complex.
- Methods have been developed, validated and published for detection or quantification of almost every metal in the periodic table using EDTA complexation.
Specifications
Liquid | |
HDPE Bottle GL45 | |
Accuracy ± 0.1% |
0.02 M (0.04 N) | |
1 L | |
Titration |