Learn More
Calcium boride, 99.5% (metals basis), Thermo Scientific Chemicals
£47.13 - £373.86
Chemical Identifiers
CAS | 12007-99-7 |
---|---|
Molecular Formula | B6Ca |
Molecular Weight (g/mol) | 104.938 |
MDL Number | MFCD00148904 |
InChI Key | AWVIDOFQNLOVRC-UHFFFAOYSA-N |
Synonym | calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide |
PubChem CID | 16212529 |
SMILES | B12B3[B-]14B5[B-]23B45.[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308878
|
Thermo Scientific Alfa Aesar
041722.06 |
5 g |
£47.13
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398868
|
Thermo Scientific Alfa Aesar
041722.14 |
25 g |
£135.68
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318878
|
Thermo Scientific Alfa Aesar
041722.22 |
100 g |
£373.86
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium boride is used as an electrode (cathode) material and n-type thermoelectric materials. It is used to make boron- alloyed steel, as a deoxidation agent in the production of oxygen-free copper. It is further used as an antioxidant in carbon bonded refractories. It acts as a new material which is used in the nuclear-industry for neutron- preventing, high-purity metal borid (TiB2,ZrB2,HfB2) and high-purity boron alloy(Ni-B, Co-B, Cu-b).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12007-99-7 | |
104.938 | |
AWVIDOFQNLOVRC-UHFFFAOYSA-N | |
16212529 |
B6Ca | |
MFCD00148904 | |
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide | |
B12B3[B-]14B5[B-]23B45.[Ca+2] |
Specifications
2235°C | |
Powder | |
MFCD00148904 | |
Insoluble in water. | |
B12B3[B-]14B5[B-]23B45.[Ca+2] | |
16212529 | |
99.5% | |
Calcium boride |
12007-99-7 | |
B6Ca | |
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide | |
AWVIDOFQNLOVRC-UHFFFAOYSA-N | |
104.938 | |
104.95 | |
2.30 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 234-525-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only