Learn More
Invitrogen™ Human Procalcitonin, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75697
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Procalcitonin (human) Recombinant Protein, RP-75697, from Invitrogen.
RP-75697 contains a purified 114 amino acid fragment (aa3-116) corresponding to the unprocessed human procalcitonin protein with an N-terminal hexahistidine tag. RP-75697 may be used as a positive control in many immunological assays. Purity >95% by SDS-PAGE.Specifications
P01258 | |
Control, Western Blot | |
796 | |
8.0 | |
E. Coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Calcitonin | |
CALCA | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.1mg/mL | |
20mM tris HCl with 250mM NaCl, 50% glycerol and no preservative; pH 8.0 | |
Human Procalcitonin | |
10μg | |
N/A | |
RUO | |
Human Procalcitonin | |
Human | |
Hu | |
Recombinant | |
Recombinant |